t-Butyl 3-(hydroxypropoxyl)-propanoate
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 3-(3-hydroxypropoxy)propanoate
OR47891-1G
OR47891-250MG
APOSOR47891-1GEA
214
EUR
APOSOR47891-1G
APOSOR47891-250MG
t-Butyl 3-(hydroxypropoxyl)-propanoate
t-Butyl 3-(hydroxypropoxyl)-propanoate
Formel:
C₁₀H₂₀O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD30730362 CAS-Nummer: 2100306-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...