Methyl 2-bromo-5-chloroisonicotinate

Lieferant: Apollo Scientific

OR47895-1G
APOSOR47895-1GEA 109 EUR
APOSOR47895-1G
Methyl 2-bromo-5-chloroisonicotinate
Methyl 2-bromo-5-chloroisonicotinate
Formel: C₇H₅BrClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14698101
CAS-Nummer: 1214336-33-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR