2-Bromo-5-chloroisonicotinaldehyde 97%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-5-chloropyridine-4-carboxaldehyde

OR47896-1G OR47896-250MG OR47896-5G
APOSOR47896-1GEA 190 EUR
APOSOR47896-1G APOSOR47896-250MG APOSOR47896-5G
2-Bromo-5-chloroisonicotinaldehyde 97%
2-Bromo-5-chloroisonicotinaldehyde
Formel: C₆H₃BrClNO
Molecular Weight: 220.451 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907898
CAS-Nummer: 921630-14-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR