4-Methylmandelic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: Hydroxy(4-methylphenyl)acetic acid, 4-[Carboxy(hydroxy)methyl]toluene

OR4793-500MG
APOSOR4793-500MGEA 0 EUR
APOSOR4793-500MG
4-Methylmandelic acid
4-Methylmandelic acid
Formel: C₉H₁₀O₃
Molecular Weight: 166.174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00961860
CAS-Nummer: 18584-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR