4-(Brommethyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Brommethyl)benzolboronsäure, (4-(Brommethyl)phenyl)metaborsäure, Dihydroxy(4-(brommethyl)phenyl)boran

OR4799-1G OR4799-5G OR4799-25G
APOSOR4799-1GEA 21.9 EUR
APOSOR4799-1G APOSOR4799-5G APOSOR4799-25G
4-(Brommethyl)phenylboronsäure
4-(Brommethyl)phenylboronsäure
Formel: C₇H₈BBrO₂
Lagertemperatur: Kühlschrank
MDL: MFCD01318113
CAS-Nummer: 68162-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR