1H-Indazole-6-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR48000-500MG
APOSOR48000-500MGEA 48 EUR
APOSOR48000-500MG
1H-Indazole-6-carbonitrile
1H-Indazole-6-carbonitrile
Formel: C₈H₅N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04973398
CAS-Nummer: 141290-59-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR