Ethyl 5-amino-4-chloro-1-ethyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480000-1G
APOSOR480000-1GEA 1550 EUR
APOSOR480000-1G
Ethyl 5-amino-4-chloro-1-ethyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-ethyl-pyrazole-3-carboxylate
Formel: C₈H₁₂ClN₃O₂
Molecular Weight: 217,653 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135133
CAS-Nummer: 1427021-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR