Ethyl 5-amino-4-bromo-1-ethyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480001-1G
APOSOR480001-1GEA 1550 EUR
APOSOR480001-1G
Ethyl 5-amino-4-bromo-1-ethyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-ethyl-pyrazole-3-carboxylate
Formel: C₈H₁₂BrN₃O₂
Molecular Weight: 262.104 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135134
CAS-Nummer: 1427012-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR