Ethyl 5-amino-1-ethyl-4-iodo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480002-1G
APOSOR480002-1GEA 1550 EUR
APOSOR480002-1G
Ethyl 5-amino-1-ethyl-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-ethyl-4-iodo-pyrazole-3-carboxylate
Formel: C₈H₁₂IN₃O₂
Molecular Weight: 309,104 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135135
CAS-Nummer: 1427013-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR