Ethyl 5-amino-4-bromo-1-isopropyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480003-1G
APOSOR480003-1GEA 1550 EUR
APOSOR480003-1G
Ethyl 5-amino-4-bromo-1-isopropyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-isopropyl-pyrazole-3-carboxylate
Formel: C₉H₁₄BrN₃O₂
Molecular Weight: 276.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135148
CAS-Nummer: 1427013-79-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR