Ethyl 5-amino-4-iodo-1-isopropyl-pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480004-1G
APOSOR480004-1GEA
1550
EUR
APOSOR480004-1G
Ethyl 5-amino-4-iodo-1-isopropyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-isopropyl-pyrazole-3-carboxylate
Formel:
C₉H₁₄IN₃O₂ Molecular Weight: 323,131 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135159 CAS-Nummer: 1427011-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...