Ethyl 5-amino-1-tert-butyl-4-chloro-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480005-1G
APOSOR480005-1GEA 1200 EUR
APOSOR480005-1G
Ethyl 5-amino-1-tert-butyl-4-chloro-pyrazole-3-carboxylate
Ethyl 5-amino-1-tert-butyl-4-chloro-pyrazole-3-carboxylate
Formel: C₁₀H₁₆ClN₃O₂
Molecular Weight: 245,706 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135171
CAS-Nummer: 1427012-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR