Ethyl 5-amino-1-tert-butyl-4-iodo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480006-1G
APOSOR480006-1GEA 912 EUR
APOSOR480006-1G
Ethyl 5-amino-1-tert-butyl-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-tert-butyl-4-iodo-pyrazole-3-carboxylate
Formel: C₁₀H₁₆IN₃O₂
Molecular Weight: 337,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135172
CAS-Nummer: 1427013-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR