Ethyl 5-amino-4-chloro-1-propyl-pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480007-1G
APOSOR480007-1GEA
1550
EUR
APOSOR480007-1G
Ethyl 5-amino-4-chloro-1-propyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-propyl-pyrazole-3-carboxylate
Formel:
C₉H₁₄ClN₃O₂ Molecular Weight: 231,679 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135173 CAS-Nummer: 1427011-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...