Ethyl 5-amino-4-bromo-1-propyl-pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480008-1G
APOSOR480008-1GEA
1550
EUR
APOSOR480008-1G
Ethyl 5-amino-4-bromo-1-propyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-propyl-pyrazole-3-carboxylate
Formel:
C₉H₁₄BrN₃O₂ Molecular Weight: 276,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135174 CAS-Nummer: 1427024-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...