Ethyl 5-amino-4-iodo-1-propyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480009-1G
APOSOR480009-1GEA 1560 EUR
APOSOR480009-1G
Ethyl 5-amino-4-iodo-1-propyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-propyl-pyrazole-3-carboxylate
Formel: C₉H₁₄IN₃O₂
Molecular Weight: 323,131 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135175
CAS-Nummer: 1427020-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR