Ethyl 5-amino-1-(2-ethoxyethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480010-1G
APOSOR480010-1GEA 1660 EUR
APOSOR480010-1G
Ethyl 5-amino-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Formel: C₁₀H₁₇N₃O₃
Molecular Weight: 227,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135176
CAS-Nummer: 1427021-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR