Ethyl 5-amino-4-chloro-1-(2-ethoxyethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480011-1G
APOSOR480011-1GEA 1770 EUR
APOSOR480011-1G
Ethyl 5-amino-4-chloro-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Formel: C₁₀H₁₆ClN₃O₃
Molecular Weight: 261,705 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135177
CAS-Nummer: 1427010-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR