Ethyl 5-amino-4-bromo-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480012-1G
APOSOR480012-1GEA
1770
EUR
APOSOR480012-1G
Ethyl 5-amino-4-bromo-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Formel:
C₁₀H₁₆BrN₃O₃ Molecular Weight: 306,156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135178 CAS-Nummer: 1427013-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...