Ethyl 5-amino-1-(2-ethoxyethyl)-4-iodo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480013-1G
APOSOR480013-1GEA 1770 EUR
APOSOR480013-1G
Ethyl 5-amino-1-(2-ethoxyethyl)-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-(2-ethoxyethyl)-4-iodo-pyrazole-3-carboxylate
Formel: C₁₀H₁₆IN₃O₃
Molecular Weight: 353,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135179
CAS-Nummer: 1427023-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR