Ethyl 5-amino-4-chloro-1-sec-butyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480014-1G
APOSOR480014-1GEA 1200 EUR
APOSOR480014-1G
Ethyl 5-amino-4-chloro-1-sec-butyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-sec-butyl-pyrazole-3-carboxylate
Formel: C₁₀H₁₆ClN₃O₂
Molecular Weight: 245,706 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135180
CAS-Nummer: 1427024-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR