Ethyl 5-amino-4-bromo-1-sec-butyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480015-1G
APOSOR480015-1GEA 2190 EUR
APOSOR480015-1G
Ethyl 5-amino-4-bromo-1-sec-butyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-sec-butyl-pyrazole-3-carboxylate
Formel: C₁₀H₁₆BrN₃O₂
Molecular Weight: 290,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135181
CAS-Nummer: 1427013-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR