Ethyl 5-amino-4-iodo-1-sec-butyl-pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480016-1G
APOSOR480016-1GEA
1200
EUR
APOSOR480016-1G
Ethyl 5-amino-4-iodo-1-sec-butyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-sec-butyl-pyrazole-3-carboxylate
Formel:
C₁₀H₁₆IN₃O₂ Molecular Weight: 337,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135182 CAS-Nummer: 1427021-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...