Ethyl 5-amino-1-(1-ethylpropyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480017-1G
APOSOR480017-1GEA 1140 EUR
APOSOR480017-1G
Ethyl 5-amino-1-(1-ethylpropyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(1-ethylpropyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₉N₃O₂
Molecular Weight: 225.287 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135183
CAS-Nummer: 1427013-80-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR