Ethyl 5-amino-4-chloro-1-(1-ethylpropyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480018-1G
APOSOR480018-1GEA
1200
EUR
APOSOR480018-1G
Ethyl 5-amino-4-chloro-1-(1-ethylpropyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(1-ethylpropyl)pyrazole-3-carboxylate
Formel:
C₁₁H₁₈ClN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135184 CAS-Nummer: 1427012-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...