Ethyl 5-amino-4-bromo-1-(1-ethylpropyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480019-1G
APOSOR480019-1GEA 1200 EUR
APOSOR480019-1G
Ethyl 5-amino-4-bromo-1-(1-ethylpropyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(1-ethylpropyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₈BrN₃O₂
Molecular Weight: 304.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135185
CAS-Nummer: 1427024-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR