Ethyl 5-amino-1-(1-ethylpropyl)-4-iodo-pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480020-1G
APOSOR480020-1GEA
1200
EUR
APOSOR480020-1G
Ethyl 5-amino-1-(1-ethylpropyl)-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-(1-ethylpropyl)-4-iodo-pyrazole-3-carboxylate
Formel:
C₁₁H₁₈IN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135186 CAS-Nummer: 1427020-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...