Ethyl 5-amino-4-iodo-1-isobutyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480023-1G
APOSOR480023-1GEA 1320 EUR
APOSOR480023-1G
Ethyl 5-amino-4-iodo-1-isobutyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-isobutyl-pyrazole-3-carboxylate
Formel: C₁₀H₁₆IN₃O₂
Molecular Weight: 337,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135189
CAS-Nummer: 1427023-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR