Ethyl 5-amino-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480024-1G
APOSOR480024-1GEA 1370 EUR
APOSOR480024-1G
Ethyl 5-amino-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₉N₃O₂
Molecular Weight: 225.287 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135190
CAS-Nummer: 1427023-15-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR