Ethyl 5-amino-4-chloro-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480025-1G
APOSOR480025-1GEA 1430 EUR
APOSOR480025-1G
Ethyl 5-amino-4-chloro-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₈ClN₃O₂
Molecular Weight: 259,733 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135191
CAS-Nummer: 1427013-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR