Ethyl 5-amino-1-(2,2-dimethylpropyl)-4-iodo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480027-1G
APOSOR480027-1GEA 1430 EUR
APOSOR480027-1G
Ethyl 5-amino-1-(2,2-dimethylpropyl)-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-(2,2-dimethylpropyl)-4-iodo-pyrazole-3-carboxylate
Formel: C₁₁H₁₈IN₃O₂
Molecular Weight: 351,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135193
CAS-Nummer: 1427021-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR