Ethyl 5-amino-1-(2-cyanoethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480028-1G
APOSOR480028-1GEA 1550 EUR
APOSOR480028-1G
Ethyl 5-amino-1-(2-cyanoethyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(2-cyanoethyl)pyrazole-3-carboxylate
Formel: C₉H₁₂N₄O₂
Molecular Weight: 208,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22107812
CAS-Nummer: 1427011-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR