Ethyl 5-amino-4-chloro-1-(2-cyanoethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480029-1G
APOSOR480029-1GEA 1610 EUR
APOSOR480029-1G
Ethyl 5-amino-4-chloro-1-(2-cyanoethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(2-cyanoethyl)pyrazole-3-carboxylate
Formel: C₉H₁₁ClN₄O₂
Molecular Weight: 242,662 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135195
CAS-Nummer: 1427012-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR