Ethyl 5-amino-4-bromo-1-(2-cyanoethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480030-1G
APOSOR480030-1GEA 1610 EUR
APOSOR480030-1G
Ethyl 5-amino-4-bromo-1-(2-cyanoethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(2-cyanoethyl)pyrazole-3-carboxylate
Formel: C₉H₁₁BrN₄O₂
Molecular Weight: 287.113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135196
CAS-Nummer: 1427023-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR