Ethyl 5-amino-1-(2-cyanoethyl)-4-iodo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480031-1G
APOSOR480031-1GEA 1610 EUR
APOSOR480031-1G
Ethyl 5-amino-1-(2-cyanoethyl)-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-(2-cyanoethyl)-4-iodo-pyrazole-3-carboxylate
Formel: C₉H₁₁IN₄O₂
Molecular Weight: 334,114 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135197
CAS-Nummer: 1427014-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR