Ethyl 5-amino-1-(cyclopropylmethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480032-1G
APOSOR480032-1GEA 912 EUR
APOSOR480032-1G
Ethyl 5-amino-1-(cyclopropylmethyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(cyclopropylmethyl)pyrazole-3-carboxylate
Formel: C₁₀H₁₅N₃O₂
Molecular Weight: 209,245 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135198
CAS-Nummer: 1427022-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR