Ethyl 5-amino-4-chloro-1-cyclohexyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480035-1G
APOSOR480035-1GEA 912 EUR
APOSOR480035-1G
Ethyl 5-amino-4-chloro-1-cyclohexyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-cyclohexyl-pyrazole-3-carboxylate
Formel: C₁₂H₁₈ClN₃O₂
Molecular Weight: 271,743 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135202
CAS-Nummer: 1427022-35-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR