Ethyl 5-amino-4-bromo-1-cyclohexyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480036-1G
APOSOR480036-1GEA 912 EUR
APOSOR480036-1G
Ethyl 5-amino-4-bromo-1-cyclohexyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-cyclohexyl-pyrazole-3-carboxylate
Formel: C₁₂H₁₈BrN₃O₂
Molecular Weight: 316,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135203
CAS-Nummer: 1427021-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR