Ethyl 5-amino-1-cyclohexyl-4-iodo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480037-1G
APOSOR480037-1GEA 1030 EUR
APOSOR480037-1G
Ethyl 5-amino-1-cyclohexyl-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-cyclohexyl-4-iodo-pyrazole-3-carboxylate
Formel: C₁₂H₁₈IN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23135204
CAS-Nummer: 1427021-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR