Ethyl 5-amino-1-tetrahydropyran-4-yl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480039-1G
APOSOR480039-1GEA 1140 EUR
APOSOR480039-1G
Ethyl 5-amino-1-tetrahydropyran-4-yl-pyrazole-3-carboxylate
Ethyl 5-amino-1-tetrahydropyran-4-yl-pyrazole-3-carboxylate
Formel: C₁₁H₁₇N₃O₃
Molecular Weight: 239,271 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135210
CAS-Nummer: 1427013-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR