Ethyl 5-amino-4-chloro-1-phenyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480041-1G
APOSOR480041-1GEA 797 EUR
APOSOR480041-1G
Ethyl 5-amino-4-chloro-1-phenyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-phenyl-pyrazole-3-carboxylate
Formel: C₁₂H₁₂ClN₃O₂
Molecular Weight: 265,696 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135212
CAS-Nummer: 1427012-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR