Ethyl 5-amino-4-iodo-1-phenyl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480042-1G
APOSOR480042-1GEA 888 EUR
APOSOR480042-1G
Ethyl 5-amino-4-iodo-1-phenyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-phenyl-pyrazole-3-carboxylate
Formel: C₁₂H₁₂IN₃O₂
Molecular Weight: 357,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135213
CAS-Nummer: 1427011-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR