Ethyl 5-amino-1-benzyl-4-chloro-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480043-1G
APOSOR480043-1GEA 797 EUR
APOSOR480043-1G
Ethyl 5-amino-1-benzyl-4-chloro-pyrazole-3-carboxylate
Ethyl 5-amino-1-benzyl-4-chloro-pyrazole-3-carboxylate
Formel: C₁₃H₁₄ClN₃O₂
Molecular Weight: 279,722 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135214
CAS-Nummer: 1427010-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR