Ethyl 5-amino-1-benzyl-4-bromo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480044-1G
APOSOR480044-1GEA 797 EUR
APOSOR480044-1G
Ethyl 5-amino-1-benzyl-4-bromo-pyrazole-3-carboxylate
Ethyl 5-amino-1-benzyl-4-bromo-pyrazole-3-carboxylate
Formel: C₁₃H₁₄BrN₃O₂
Molecular Weight: 324.173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135215
CAS-Nummer: 1427023-40-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR