Ethyl 5-amino-1-benzyl-4-iodo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480045-1G
APOSOR480045-1GEA 912 EUR
APOSOR480045-1G
Ethyl 5-amino-1-benzyl-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-benzyl-4-iodo-pyrazole-3-carboxylate
Formel: C₁₃H₁₄IN₃O₂
Molecular Weight: 371,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135216
CAS-Nummer: 1427014-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR