Ethyl 5-amino-4-chloro-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480046-1G
APOSOR480046-1GEA
797
EUR
APOSOR480046-1G
Ethyl 5-amino-4-chloro-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Formel:
C₁₃H₁₄ClN₃O₃ Molecular Weight: 295,722 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135217 CAS-Nummer: 1427014-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...