Ethyl 5-amino-4-bromo-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480047-1G
APOSOR480047-1GEA
797
EUR
APOSOR480047-1G
Ethyl 5-amino-4-bromo-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Formel:
C₁₃H₁₄BrN₃O₃ Molecular Weight: 340,173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135218 CAS-Nummer: 1427011-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...