Ethyl 5-amino-4-iodo-1-(4-methoxyphenyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480048-1G
APOSOR480048-1GEA 912 EUR
APOSOR480048-1G
Ethyl 5-amino-4-iodo-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Formel: C₁₃H₁₄IN₃O₃
Molecular Weight: 387,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135219
CAS-Nummer: 1427013-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR