Ethyl 5-amino-1-(3-methoxyphenyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480049-1G
APOSOR480049-1GEA 797 EUR
APOSOR480049-1G
Ethyl 5-amino-1-(3-methoxyphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(3-methoxyphenyl)pyrazole-3-carboxylate
Formel: C₁₃H₁₅N₃O₃
Molecular Weight: 261,277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07388422
CAS-Nummer: 1427013-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR