Ethyl 5-amino-4-chloro-1-(p-tolyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480051-1G
APOSOR480051-1GEA
1090
EUR
APOSOR480051-1G
Ethyl 5-amino-4-chloro-1-(p-tolyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(p-tolyl)pyrazole-3-carboxylate
Formel:
C₁₃H₁₄ClN₃O₂ Molecular Weight: 279,722 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135220 CAS-Nummer: 1427021-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...